missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DRA Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DRA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8909-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_061984.2 | |
HLA-DRA Protein | |
Research Use Only | |
HLA-DRA | |
Recombinant Protein | |
E. coli | |
ILIVDLDVHH GNGTQQAFYA DPSILYISLH RYDEGNFFPG SGAPNEVGTG LGEGYNINIA WTGGLDPPMG DVEYLEAFRT IVKPVAKEFD PDMVLVSAGF DALEGHTPPL GGYKVTAKCF GHLTKQLMTL ADGRVVLALE GGHDLTAICD ASEACVNALL GNELEPLAED ILHQSPNMNA VISLQKIIEI QSMSLKFS | |
Greater than 90% as determined by SDS-PAGE. |
3122 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |