missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DRA Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DRA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8909-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017131

  • 530.00€ / 100µg
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_061984.2
HLA-DRA Protein
Research Use Only
HLA-DRA
Recombinant Protein
E. coli
ILIVDLDVHH GNGTQQAFYA DPSILYISLH RYDEGNFFPG SGAPNEVGTG LGEGYNINIA WTGGLDPPMG DVEYLEAFRT IVKPVAKEFD PDMVLVSAGF DALEGHTPPL GGYKVTAKCF GHLTKQLMTL ADGRVVLALE GGHDLTAICD ASEACVNALL GNELEPLAED ILHQSPNMNA VISLQKIIEI QSMSLKFS
Greater than 90% as determined by SDS-PAGE.
3122
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers