missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DPB1 Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DPB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8814-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046631

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_002112.3
HLA-DPB1 Protein
Research Use Only
HLA-DPB1
Recombinant Protein
E. coli
RATPENYLFQ GRQECYAFNG TQRFLERYIY NREEFARFDS DVGEFRAVTE LGRPAAEYWN SQKDILEEKR AVPDRMCRHN YELGGPMTLQ RRVQPRVNVS PSKKGPLQHH NLLVCHVTDF YPGSIQVRWF LNGQEETAGV VSTNLIRNGD WTFQILVMLE MTPQQGDVYT CQVEHTSLDS PVTVEWKAQS DSARSK
Greater than 90% as determined by SDS-PAGE.
3115
500 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers