missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2B type 1-N Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-N, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8012-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003511.1 | |
Histone H2B type 1-N Protein | |
Research Use Only | |
HIST1H2BN | |
Recombinant Protein | |
E. coli | |
PEPSKSAPAP KKGSKKAVTK AQKKDGKKRK RSRKESYSVY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSSK | |
Greater than 90% as determined by SDS-PAGE. |
8341 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |