missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6151-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129513.1 | |
Histone deacetylase 11 Protein | |
Research Use Only | |
HDAC11 | |
Recombinant Protein | |
E. coli | |
MLHTTQLYQH VPETRWPIVY SPRYNITFMG LEKLHPFDAG KWGKVINFLK EEKLLSDSML VEAREASEED LLVVHTRRYL NELKWSFAVA TITEIPPVIF LPNFLVQRKV LRPLRTQTGG TIMAGKLAVE RGWAINVGGG FHHCSSDRGG GFCAYADITL AIKFLFERVE GISRATIIDL DAHQGNGHER DFMDDKRVYI MDVYNRHIYP GDRFAKQAIR RKVELEWGTE DDEYLDKVER NIKKSLQEHL PDVVVYNAGT DILEGDRLGG LSISPAGIVK RDELVFRMVR GRRVPILMVT SGGYQKRTAR IIADSILNLF GLGLIGPESP SVSAQNSDTP LLPPAVP | |
Greater than 80% as determined by SDS-PAGE. |
79885 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |