missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heme oxygenase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Heme oxygenase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6172-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002124.1 | |
Heme oxygenase 1 Protein | |
Research Use Only | |
HMOX1 | |
Recombinant Protein | |
E. coli | |
RPQPDSMPQD LSEALKEATK EVHTQAENAE FMRNFQKGQV TRDGFKLVMA SLYHIYVALE EEIERNKESP VFAPVYFPEE LHRKAALEQD LAFWYGPRWQ EVIPYTPAMQ RYVKRLHEVG RTEPELLVAH AYTRYLGDLS GGQVLKKIAQ KALDLPSSGE GLAFFTFPNI ASATKFKQLY RSRMNSLEMT PAVRQRVIEE AKTAFLLNIQ LFEELQELLT HDTKDQSPSR APGLRQRASN KVQDSAPVET PRGKPPLNTR SQAPLLRWVL TLSFLVATVA VGLYAM | |
Greater than 90% as determined by SDS-PAGE. |
3162 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |