missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heme oxygenase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Heme oxygenase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6172-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051320

  • 2385.00€ / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_002124.1
Heme oxygenase 1 Protein
Research Use Only
HMOX1
Recombinant Protein
E. coli
RPQPDSMPQD LSEALKEATK EVHTQAENAE FMRNFQKGQV TRDGFKLVMA SLYHIYVALE EEIERNKESP VFAPVYFPEE LHRKAALEQD LAFWYGPRWQ EVIPYTPAMQ RYVKRLHEVG RTEPELLVAH AYTRYLGDLS GGQVLKKIAQ KALDLPSSGE GLAFFTFPNI ASATKFKQLY RSRMNSLEMT PAVRQRVIEE AKTAFLLNIQ LFEELQELLT HDTKDQSPSR APGLRQRASN KVQDSAPVET PRGKPPLNTR SQAPLLRWVL TLSFLVATVA VGLYAM
Greater than 90% as determined by SDS-PAGE.
3162
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers