missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heme oxygenase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Heme oxygenase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6172-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16041320

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_002124.1
Heme oxygenase 1 Protein
Research Use Only
HMOX1
Recombinant Protein
E. coli
RPQPDSMPQD LSEALKEATK EVHTQAENAE FMRNFQKGQV TRDGFKLVMA SLYHIYVALE EEIERNKESP VFAPVYFPEE LHRKAALEQD LAFWYGPRWQ EVIPYTPAMQ RYVKRLHEVG RTEPELLVAH AYTRYLGDLS GGQVLKKIAQ KALDLPSSGE GLAFFTFPNI ASATKFKQLY RSRMNSLEMT PAVRQRVIEE AKTAFLLNIQ LFEELQELLT HDTKDQSPSR APGLRQRASN KVQDSAPVET PRGKPPLNTR SQAPLLRWVL TLSFLVATVA VGLYAM
Greater than 90% as determined by SDS-PAGE.
3162
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers