missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GRK2 / ADRBK1 Protein
A DNA sequence encoding the Homo sapiens (Human) GRK2 / ADRBK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8340-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001610.2 | |
GRK2 / ADRBK1 Protein | |
Research Use Only | |
ADRBK1 | |
Recombinant Protein | |
E. coli | |
ADLEAVLADV SYLMAMEKSK ATPAARASKK ILLPEPSIRS VMQKYLEDRG EVTFEKIFSQ KLGYLLFRDF CLNHLEEARP LVEFYEEIKK YEKLETEEER VARSREIFDS YIMKELLACS HPFSKSATEH VQGHLGKKQV PPDLFQPYIE EICQNLRGDV FQKFIESDKF TRFCQWKNVE LNIHLTMNDF SVHRIIGRGG FGEVYGCRKA DTGKMYAMKC | |
Greater than 90% as determined by SDS-PAGE. |
156 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |