missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human gp100 / PMEL17 / SILV Protein

A DNA sequence encoding the Homo sapiens (Human) gp100 / PMEL17 / SILV, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6683-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035080

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001186982.1
gp100 / PMEL17 / SILV Protein
Research Use Only
SILV
Recombinant Protein
Yeast
KVPRNQDWLG VSRQLRTKAW NRQLYPEWTE AQRLDCWRGG QVSLKVSNDG PTLIGANASF SIALNFPGSQ KVLPDGQVIW VNNTIINGSQ VWGGQPVYPQ ETDDACIFPD GGPCPSGSWS QKRSFVYVWK TWGQYWQVLG GPVSGLSIGT GRAMLGTHTM EVTVYHRRGS RSYVPLAHSS SAFTITDQVP FSVSVSQLRA LDGGNKHFLR NQPLTFALQL HDPSGYLAEA DLSYTWDFGD SSGTLISRAL VVTHTYLEPG PVTAQVVLQA AIPLTSCGSS PVPGTTDGHR PTAEAPNTTA GQVPTTEVVG TTPGQAPTAE PSGTTSVQVP TTEVISTAPV QMPTAESTGM TPEKVPVSEV MGTTLAEMST PEATGMTPAE VSIVVLSGTT AAQVTTTEWV ETTARELPIP EPEGPDASSI MSTESITGSL GPLLDGTATL RLV
Greater than 90% as determined by SDS-PAGE.
6490
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers