missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNL3L Protein
A DNA sequence encoding the Homo sapiens (Human) GNL3L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8075-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171748.1 | |
GNL3L Protein | |
Research Use Only | |
GNL3L | |
Recombinant Protein | |
E. coli | |
MMKLRHKNKK PGEGSKGHKK ISWPYPQPAK QNGKKATSKV PSAPHFVHPN DHANREAELK KKWVEEMREK QQAAREQERQ KRRTIESYCQ DVLRRQEEFE HKEEVLQELN MFPQLDDEAT RKAYYKEFRK VVEYSDVILE VLDARDPLGC RCFQMEEAVL RAQGNKKLVL VLNKIDLVPK EVVEKWLDYL RNELPTVAFK ASTQHQVKNL NRCSVPVDQA SESLLKSKAC FGAENLMRVL GNYCRLGEVR THIRVGVVGL PNVGKSSLIN SLKRSRACSV GAVPGITKFM QEVYLDKFIR LLDAPGIVPG PNSEVGTILR NCVHVQKLAD PVTPVETILQ RCNLEEISNY YGVSGFQTTE HFLTAVAHRL GKKKKGGLYS QEQAAKAVLA DWVSGKISFY IPPPATHTLP THLSAEIVKE MTEVFDIEDT EQANEDTMEC LATGESDELL GDTDPLEMEI KLLHSPMTKI ADAIENKTTV YKIGDLTGYC TNPNRHQMGW AKRNVDHRPK SNSMVDVCSV DRRSVLQRIM ETDPLQQGQA LASALKNKKK MQKRADKIAS KLSDSMMSAL DLSGNADDGV GD | |
Greater than 90% as determined by SDS-PAGE. |
54552 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |