missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glucosamine-6-phosphate isomerase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Glucosamine-6-phosphate isomerase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6104-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040830

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_005462.1
Glucosamine-6-phosphate isomerase 1 Protein
Research Use Only
GNPDA1
Recombinant Protein
E. coli
MKLIILEHYS QASEWAAKYI RNRIIQFNPG PEKYFTLGLP TGSTPLGCYK KLIEYYKNGD LSFKYVKTFN MDEYVGLPRD HPESYHSFMW NNFFKHIDIH PENTHILDGN AVDLQAECDA FEEKIKAAGG IELFVGGIGP DGHIAFNEPG SSLVSRTRVK TLAMDTILAN ARFFDGELTK VPTMALTVGV GTVMDAREVM ILITGAHKAF ALYKAIEEGV NHMWTVSAFQ QHPRTVFVCD EDATLELKVK TVKYFKGLML VHNKLVDPLY SIKEKETEKS QSSKKPYSD
Greater than 90% as determined by SDS-PAGE.
10007
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers