missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GIGYF2 Protein
A DNA sequence encoding the Homo sapiens (Human) GIGYF2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9814-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001096616.1 | |
GIGYF2 Protein | |
Research Use Only | |
GIGYF2 | |
Recombinant Protein | |
Yeast | |
ERLASKLQEH RAKGVSIPLM HEAMQKWYYK DPQGEIQGPF NNQEMAEWFQ AGYFTMSLLV KRACDESFQP LGDIMKMWGR VPFSPGPAPP PHMGELDQER LTRQQELTAL YQMQHLQYQQ FLIQQQYAQV LAQQQKAALS SQQQQQLALL LQQFQTLKMR ISDQNIIPSV TRSVSVPDTG SIWELQPTAS QPTVWEGGSV WDLPLDTTTP GPALEQLQQL EKAKAAKLEQ ERREAEMRAK REEEERKRQE ELRRQQEEIL RRQQEEERKR REEEELARRK QEEALRRQRE QEIALRRQRE EEERQQQEEA LRRLEERRRE EEERRKQEEL LRKQEEEAAK WAREEEEAQR RLEENRLRME EEAARLRHEE EERKRKELEV QRQKELMRQR QQQQEALRRL QQQQQQQQLA QMKLPSSSTW GQQSNTTACQ SQATLSLAEI QKLEEERERQ LREEQRRQQR ELMKALQQQQ QQQQQKLSGW GNVSKPSGTT KSLLEIQQEE ARQMQKQQQQ QQQHQQPNRA R | |
Greater than 90% as determined by SDS-PAGE. |
26058 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |