missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galectin-1 / LGALS1 Protein
A DNA sequence encoding the Homo sapiens (Human) Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6294-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002296.1 | |
Galectin-1 / LGALS1 Protein | |
Research Use Only | |
LGALS1 | |
Recombinant Protein | |
E. coli | |
ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD | |
Greater than 90% as determined by SDS-PAGE. |
3956 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |