missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human G-protein coupled receptor 161 Protein

A DNA sequence encoding the Homo sapiens (Human) G-protein coupled receptor 161, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8467-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064771

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001254538.1
G-protein coupled receptor 161 Protein
Research Use Only
GPR161
Recombinant Protein
E. coli
MSLNSSLSCR KELSNLTEEE GGEGGVII
Greater than 90% as determined by SDS-PAGE.
23432
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers