missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human G-protein coupled receptor 161 Protein

A DNA sequence encoding the Homo sapiens (Human) G-protein coupled receptor 161, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8467-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044771

  • 2385.00€ / 1mg
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001254538.1
G-protein coupled receptor 161 Protein
Research Use Only
GPR161
Recombinant Protein
E. coli
MSLNSSLSCR KELSNLTEEE GGEGGVII
Greater than 90% as determined by SDS-PAGE.
23432
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers