missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroblast growth factor 5 Protein

A DNA sequence encoding the Homo sapiens (Human) Fibroblast growth factor 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8665-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055851

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_004455.2
Fibroblast growth factor 5 Protein
Research Use Only
FGF5
Recombinant Protein
E. coli
AWAHGEKRLA PKGQPGPAAT DRNPRGSSSR QSSSSAMSSS SASSSPAASL GSQGSGLEQS SFQWSPSGRR TGSLYCRVGI GFHLQIYPDG KVNGSHEANM LSVLEIFAVS QGIVGIRGVF SNKFLAMSKK GKLHASAKFT DDCKFRERFQ ENSYNTYASA IHRTEKTGRE WYVALNKRGK AKRGCSPRVK PQHISTHFLP RFKQSEQPEL SFTVTVPEKK KPPSPIKPKI PLSAPRKNTN SVKYRLKFRF G
Greater than 90% as determined by SDS-PAGE.
2250
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers