missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ES1 protein homolog, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ES1 protein homolog, mitochondrial, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_004640.4 |
---|---|
Gene ID (Entrez) | 102724023 |
Name | ES1 protein homolog, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024351
|
enQuireBio™
QP8386-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16044351
|
enQuireBio™
QP8386-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16034351
|
enQuireBio™
QP8386-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054351
|
enQuireBio™
QP8386-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064351
|
enQuireBio™
QP8386-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074351
|
enQuireBio™
QP8386-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_004640.4 | |
ES1 protein homolog, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
102724023 | |
Research Use Only | |
C21orf33 | |
Recombinant Protein | |
E. coli | |
ARVALVLSGC GVYDGTEIHE ASAILVHLSR GGAEVQIFAP DVPQMHVIDH TKGQPSEGES RNVLTESARI ARGKITDLAN LSAANHDAAI FPGGFGAAKN LSTFAVDGKD CKVNKEVERV LKEFHQAGKP IGLCCIAPVL AAKVLRGVEV TVGHEQEEGG KWPYAGTAEA IKALGAKHCV KEVVEAHVDQ KNKVVTTPAF MCETALHYIH DGIGAMVRKV LELTGK | |
Greater than 90% as determined by SDS-PAGE. |