missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ER membrane protein complex subunit 9 Protein
A DNA sequence encoding the Homo sapiens (Human) ER membrane protein complex subunit 9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8297-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057133.2 | |
ER membrane protein complex subunit 9 Protein | |
Research Use Only | |
EMC9 | |
Recombinant Protein | |
E. coli | |
MGEVEISALA YVKMCLHAAR YPHAAVNGLF LAPAPRSGEC LCLTDCVPLF HSHLALSVML EVALNQVDVW GAQAGLVVAG YYHANAAVND QSPGPLALKI AGRIAEFFPD AVLIMLDNQK LVPQPRVPPV IVLENQGLRW VPKDKNLVMW RDWEESRQMV GALLEDRAHQ HLVDFDCHLD DIRQDWTNQR LNTQITQWVG PTNGNGNA | |
Greater than 90% as determined by SDS-PAGE. |
51016 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |