missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Epcr / PROCR Protein

A DNA sequence encoding the Homo sapiens (Human) Epcr / PROCR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8541-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065181

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_006395.2
Epcr / PROCR Protein
Research Use Only
PROCR
Recombinant Protein
E. coli
SQDASDGLQR LHMLQISYFR DPYHVWYQGN ASLGGHLTHV LEGPDTNTTI IQLQPLQEPE SWARTQSGLQ SYLLQFHGLV RLVHQERTLA FPLTIRCFLG CELPPEGSRA HVFFEVAVNG SSFVSFRPER ALWQADTQVT SGVVTFTLQQ LNAYNRTRYE LREFLEDTCV QYVQKHISAE NTKGSQTSRS YTS
Greater than 90% as determined by SDS-PAGE.
10544
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers