missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Embryonic stem cell-related gene Protein
A DNA sequence encoding the Homo sapiens (Human) Embryonic stem cell-related gene protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7626-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Embryonic stem cell-related gene Protein | |
Research Use Only | |
ESRG | |
Recombinant Protein | |
E. coli | |
MTLFSDSARL HPGEINSLVA HTKPVWWSLH TDAHEIWCRD SDRGTSLGRS IPCPPALCSV RKIHLRPQVL RPTSPRNISP ISNPVSGLFL LCSPTSLTIP QPLSPFNLGA TLQSLPSLNF NSFHSLVETK ETCFIREPKT PAPVTDWEGS LPLVFNHCRD ASLISRFRPR RDACLGPSPL AASPAFLGQG QVPLNPFSFT LSGKSRFSGA GASTPQPLLL HP | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |