missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Docking protein 1
A DNA sequence encoding the Homo sapiens (Human) Docking protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8017-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001184189.1 | |
Docking protein 1 | |
Research Use Only | |
DOK1 | |
Recombinant Protein | |
E. coli | |
MDGAVMEGPL FLQSQRFGTK RWRKTWAVLY PASPHGVARL EFFDHKGSSS GGGRGSSRRL DCKVIRLAEC VSVAPVTVET PPEPGATAFR LDTAQRSHLL AADAPSSAAW VQTLCRNAFP KGSWTLAPTD NPPKLSALEM LENSLYSPTW EGSQFWVTVQ RTEAAERCGL HGSYVLRVEA ERLTLLTVGA QSQILEPLLS WPYTLLRRYG RDKVMFSFEA GRRCPSGPGT FTFQTAQGND IFQAVETAIH RQKAQGKAGQ GHDVLRADSH EGEVAEGKLP SPPGPQELLD SPPALYAEPL DSLRIAPCPS QDSLYSDPLD STSAQAGEGV QRKKPLYWDL YEHAQQQLLK AKLTDPKEDP IYDEPEGLAP VPPQGLYDLP REPKDAWWCQ ARVKEEGYEL PYNPATDDYA VPPPRSTKPL LAPKPQGPAF PEPGTATGSG IKSHNSALYS QVQKSGASGS WDCGLSRVGT DKTGVKSEGS T | |
Greater than 90% as determined by SDS-PAGE. |
1796 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |