missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein complementing XP-C cells Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein complementing XP-C cells, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6892-ec-50ug
Detalles adicionales : Weight : 0.01000kg
Especificaciones
NP_004619.3 | |
DNA repair protein complementing XP-C cells Protein | |
Research Use Only | |
XPC | |
Recombinant Protein | |
E. coli | |
SLPAASSSSS SSKRGKKMCS DGEKAEKRSI AGIDQWLEVF CEQEEKWVCV DCVHGVVGQP LTCYKYATKP MTYVVGIDSD GWVRDVTQRY DPVWMTVTRK CRVDAEWWAE TLRPYQSPFM DREKKEDLEF QAKHMDQPLP TAIGLYKNHP LYALKRHLLK YEAIYPETAA ILGYCRGEAV YSRDCVHTLH SRDTWLKKAR VVRLGEVPYK MVKGFSNRAR KARLAEPQLR EENDLGLFG | |
Greater than 90% as determined by SDS-PAGE. |
7508 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |