missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclin-J-like Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclin-J-like protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9039-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001295102.1 | |
Cyclin-J-like Protein | |
Research Use Only | |
CCNJL | |
Recombinant Protein | |
Yeast | |
MVPGTPPTPT QVLFQPPAYP ALGQPATTLA QFQTPVQDLC LAYRDSLQAH RSGSLLSGST GSSLHTPYQP LQPLDMCPVP VPASLSMHMA IAAEPRHCLA TTYGSSYFSG SHMFPTGCFD R | |
Greater than 90% as determined by SDS-PAGE. |
79616 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |