missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CPEB1 Protein
A DNA sequence encoding the Homo sapiens (Human) CPEB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8045-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001073001.1 | |
CPEB1 Protein | |
Research Use Only | |
CPEB1 | |
Recombinant Protein | |
E. coli | |
MLFPTSAQES SRGLPDANDL CLGLQSLSLT GWDRPWSTQD SDSSAQSSTH SVLSMLHNPL GNVLGKPPLS FLPLDPLGSD LVDKFPAPSV RGSRLDTRPI LDSRSSSPSD SDTSGFSSGS DHLSDLISSL RISPPLPFLS LSGGGPRDPL KMGVGSRMDQ EQAALAAVTP SPTSASKRWP GASVWPSWDL LEAPKDPFSI EREARLHRQA AAVNEATCTW SGQLPPRNYK NPIYSCKVFL GGVPWDITEA GLVNTFRVFG SLSVEWPGKD GKHPRCPPKG YVYLVFELEK SVRSLLQACS HDPLSPDGLS EYYFKMSSRR MRCKEVQVIP WVLADSNFVR SPSQRLDPSR TVFVGALHGM LNAEALAAIL NDLFGGVVYA GIDTDKHKYP IGSGRVTFNN QRSYLKAVSA AFVEIKTTKF TKKVQIDPYL EDSLCHICSS QPGPFFCRDQ VCFKYFCRSC WHWRHSMEGL RHHSPLMRNQ KNRDSS | |
Greater than 90% as determined by SDS-PAGE. |
64506 | |
50 μg | |
Not determined. | |
Full Length of isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |