missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COX5A, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) COX5A, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8319-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013991

  • 2385.00€ / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_004246.2
COX5A, mitochondrial Protein
Research Use Only
COX5A
Recombinant Protein
E. coli
SHGSQETDEE FDARWVTYFN KPDIDAWELR KGINTLVTYD MVPEPKIIDA ALRACRRLND FASTVRILEV VKDKAGPHKE IYPYVIQELR PTLNELGIST PEELGLDKV
Greater than 90% as determined by SDS-PAGE.
9377
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers