missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMM domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) COMM domain-containing protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8487-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001271306.1 | |
COMM domain-containing protein 1 | |
Research Use Only | |
COMMD4 | |
Recombinant Protein | |
E. coli | |
MRFRFCGDLD CPDWVLAEIS TLAKMSSVKL RLLCSQVLKE LLGQGIDYEK ILKLTADAKF ESGDVKATVA VLSFILSSAA KHSVDGESLS SELQQLGLPK EHAASLCRCY EEKQSPLQKH LRVCSLRMNR LAGVGWRVDY TLSSSLLQSV EEPMVHLRLE VAAAPGTPAQ PVAMSLSADK FQVLLAELKQ AQTLM | |
Greater than 90% as determined by SDS-PAGE. |
54939 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |