missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKMT1A Protein

A DNA sequence encoding the Homo sapiens (Human) CKMT1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8676-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035911

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001015001.1
CKMT1A Protein
Research Use Only
CKMT1A
Recombinant Protein
E. coli
ASERRRLYPP SAEYPDLRKH NNCMASHLTP AVYARLCDKT TPTGWTLDQC IQTGVDNPGH PFIKTVGMVA GDEETYEVFA DLFDPVIQER HNGYDPRTMK HTTDLDASKI RSGYFDERYV LSSRVRTGRS IRGLSLPPAC TRAERREVER VVVDALSGLK GDLAGRYYRL SEMTEAEQQQ LIDDHFLFDK PVSPLLTAAG MARDWPDARG IWHNNEKSFL IWVNEEDHTR VISMEKGGNM KRVFERFCRG LKEVERLIQE RGWEFMWNER LGYILTCPSN LGTGLRAGVH IKLPLLSKDS RFPKILENLR LQKRGTGGVD TAATGGVFDI SNLDRLGKSE VELVQLVIDG VNYLIDCERR LERGQDIRIP TPVIHTKH
Greater than 90% as determined by SDS-PAGE.
1159
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers