missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cilia- and flagella-associated protein 20 Protein
A DNA sequence encoding the Homo sapiens (Human) Cilia- and flagella-associated protein 20, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8284-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037374.1 | |
Cilia- and flagella-associated protein 20 Protein | |
Research Use Only | |
C16orf80 | |
Recombinant Protein | |
E. coli | |
MFKNTFQSGF LSILYSIGSK PLQIWDKKVR NGHIKRITDN DIQSLVLEIE GTNVSTTYIT CPADPKKTLG IKLPFLVMII KNLKKYFTFE VQVLDDKNVR RRFRASNYQS TTRVKPFICT MPMRLDDGWN QIQFNLLDFT RRAYGTNYIE TLRVQIHANC RIRRVYFSDR LYSEDELPAE FKLYLPVQNK AKQ | |
Greater than 80% as determined by SDS-PAGE. |
29105 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |