missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Chitinase domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Chitinase domain-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8193-ma-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001136146.1 | |
Chitinase domain-containing protein 1 | |
Research Use Only | |
Chid1 | |
Recombinant Protein | |
Mammal | |
TLSKSDAKKA ASKTLLEKSQ FSDKPVQDRG LVVTDLKAES VVLEHRSYCS AKARDRHFAG DVLGYVTPWN SHGYDVTKVF GSKFTQISPV WLQLKRRGRE MFEVTGLHDV DQGWMRAVRK HAKGLHIVPR LLFEDWTYDD FRNVLDSEDE IEELSKTVVQ VAKNQHFDGF VVEVWNQLLS QKRVGLIHML THLAEALHQA RLLALLVIPP AITPGTDQLG MFTHKEFEQL APVLDGFSLM TYDYSTAHQP GPNAPLSWVR ACVQVLDPKS KWRSKILLGL NFYGMDYATS KDAREPVVGA RYIQTLKDHR PRMVWDSQAS EHFFEYKKSR SGRHVVFYPT LKSLQVRLEL ARELGVGVSI WELGQGLDYF YDLL | |
Greater than 90% as determined by SDS-PAGE. |
66005 | |
20 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |