missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD52 / CDW52 Protein
A DNA sequence encoding the Homo sapiens (Human) CD52 / CDW52, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9049-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001794.2 | |
CD52 / CDW52 Protein | |
Research Use Only | |
CD52 | |
Recombinant Protein | |
Yeast | |
GQNDTSQTSS PS | |
Greater than 90% as determined by SDS-PAGE. |
1043 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |