missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD45 / PTPRC Protein

A DNA sequence encoding the Homo sapiens (Human) CD45 / PTPRC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9356-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008181

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_002829.3
CD45 / PTPRC Protein
Research Use Only
PTPRC
Recombinant Protein
Yeast
QSPTPSPTGL TTAKMPSVPL SSDPLPTHTT AFSPASTFER ENDFSETTTS LSPDNTSTQV SPDSLDNASA FNTTGVSSVQ TPHLPTHADS QTPSAGTDTQ TFSGSAANAK LNPTPGSNAI SDVPGERSTA STFPTDPVSP LTTTLSLAHH SSAALPARTS NTTITANTSD AYLNASETTT LSPSGSAVIS TTTIATTPSK PTCDEKYANI TVDYLYNKET KLFTAKLNVN ENVECGNNTC TNNEVHNLTE CKNASVSISH NSCTAPDKTL ILDVPPGVEK FQLHDCTQVE KADTTICLKW KNIETFTCDT QNITYRFQCG NMIFDNKEIK LENLEPEHEY KCDSEILYNN HKFTNASKII KTDFGSPGEP QIIFCRSEAA HQGVITWNPP QRSFHNFTLC YIKETEKDCL NLDKNLIKYD LQNLKPYTKY VLSLHAYIIA KVQRNGSAAM CHFTTKSAPP SQVWNMTVSM TSDNSMHVKC RPPRDRNGPH ERYHLEVEAG NTLVRNESHK NCDFRVKDLQ YSTDYTFKAY FHNGDYPGEP FILHHSTSYN SK
Greater than 90% as determined by SDS-PAGE.
5788
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers