missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD155 / PVR Protein

A DNA sequence encoding the Homo sapiens (Human) CD155 / PVR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6559-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084150

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001129240.1
CD155 / PVR Protein
Research Use Only
PVR
Recombinant Protein
E. coli
WPPPGTGDVV VQAPTQVPGF LGDSVTLPCY LQVPNMEVTH VSQLTWARHG ESGSMAVFHQ TQGPSYSESK RLEFVAARLG AELRNASLRM FGLRVEDEGN YTCLFVTFPQ GSRSVDIWLR VLAKPQNTAE VQKVQLTGEP VPMARCVSTG GRPPAQITWH SDLGGMPNTS QVPGFLSGTV TVTSLWILVP SSQVDGKNVT CKVEHESFEK PQLLTVNLTV YYPPEVSISG YDNNWYLGQN EATLTCDARS NPEPTGYNWS TTMGPLPPFA VAQGAQLLIR PVDKPINTTL ICNVTNALGA RQAELTVQVK EGPPSEHSGI SRN
Greater than 90% as determined by SDS-PAGE.
5817
10 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers