missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CBFB / CBF-beta Protein
A DNA sequence encoding the Homo sapiens (Human) CBFB / CBF-beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7610-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001746.1 | |
CBFB / CBF-beta Protein | |
Research Use Only | |
CBFB | |
Recombinant Protein | |
E. coli | |
MPRVVPDQRS KFENEEFFRK LSRECEIKYT GFRDRPHEER QARFQNACRD GRSEIAFVAT GTNLSLQFFP ASWQGEQRQT PSREYVDLER EAGKVYLKAP MILNGVCVIW KGWIDLQRLD GMGCLEFDEE RAQQEDALAQ QAFEEARRRT REFEDRDRSH REEMEVRVSQ LLAVTGKKTT RP | |
Greater than 90% as determined by SDS-PAGE. |
865 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |