missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calmodulin Protein
A DNA sequence encoding the Homo sapiens (Human) Calmodulin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9025-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001292553.1 | |
Calmodulin Protein | |
Research Use Only | |
CALM1 | |
Recombinant Protein | |
Yeast | |
ADQLTEEQIA EFKEAFSLFD KDGDGTITTK ELGTVMRSLG QNPTEAELQD MINEVDADGN GTIDFPEFLT MMARKMKDTD SEEEIREAFR VFDKDGNGYI SAAELRHVMT NLGEKLTDEE VDEMIREADI DGDGQVNYEE FVQMMTAK | |
Greater than 90% as determined by SDS-PAGE. |
801 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |