missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALML5 / CLSP Protein
A DNA sequence encoding the Homo sapiens (Human) CALML5 / CLSP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8302-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_059118.2 | |
CALML5 / CLSP Protein | |
Research Use Only | |
CALML5 | |
Recombinant Protein | |
E. coli | |
AGELTPEEEA QYKKAFSAVD TDGNGTINAQ ELGAALKATG KNLSEAQLRK LISEVDSDGD GEISFQEFLT AAKKARAGLE DLQVAFRAFD QDGDGHITVD ELRRAMAGLG QPLPQEELDA MIREADVDQD GRVNYEEFAR MLAQE | |
Greater than 90% as determined by SDS-PAGE. |
51806 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |