missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALCA / CGRP Protein
A DNA sequence encoding the Homo sapiens (Human) CALCA / CGRP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8831-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001029125.1 | |
CALCA / CGRP Protein | |
Research Use Only | |
CALCA | |
Recombinant Protein | |
E. coli | |
ACDTATCVTH RLAGLLSRSG GVVKNNFVPT NVGSKA | |
Greater than 90% as determined by SDS-PAGE. |
796 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |