missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bile acid receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Bile acid receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7899-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061481

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001193906.1
Bile acid receptor Protein
Research Use Only
NR1H4
Recombinant Protein
E. coli
MVMQFQGLEN PIQISPHCSC TPSGFFMEMM SMKPAKGVLT EQVAGPLGQN LEVEPYSQYS NVQFPQVQPQ ISSSSYYSNL GFYPQQPEEW YSPGIYELRR MPAETLYQGE TEVAEMPVTK KPRMGASAGR IKGDELCVVC GDRASGYHYN ALTCEGCKGF FRRSITKNAV YKCKNGGNCV MDMYMRRKCQ ECRLRKCKEM GMLAECMYTG LLTEIQCKSK RLRKNVKQHA DQTVNEDSEG RDLRQVTSTT KSCREKTELT PDQQTLLHFI MDSYNKQRMP QEITNKILKE EFSAEENFLI LTEMATNHVQ VLVEFTKKLP GFQTLDHEDQ IALLKGSAVE AMFLRSAEIF NKKLPSGHSD LLEERIRNSG ISDEYITPMF SFYKSIGELK MTQEEYALLT AIVILSPDRQ YIKDREAVEK LQEPLLDVLQ KLCKIHQPEN PQHFACLLGR LTELRTFNHH HAEMLMSWRV NDHKFT
Greater than 90% as determined by SDS-PAGE.
9971
50 μg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers