missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bile acid receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Bile acid receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7899-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001193906.1 | |
Bile acid receptor Protein | |
Research Use Only | |
NR1H4 | |
Recombinant Protein | |
E. coli | |
MVMQFQGLEN PIQISPHCSC TPSGFFMEMM SMKPAKGVLT EQVAGPLGQN LEVEPYSQYS NVQFPQVQPQ ISSSSYYSNL GFYPQQPEEW YSPGIYELRR MPAETLYQGE TEVAEMPVTK KPRMGASAGR IKGDELCVVC GDRASGYHYN ALTCEGCKGF FRRSITKNAV YKCKNGGNCV MDMYMRRKCQ ECRLRKCKEM GMLAECMYTG LLTEIQCKSK RLRKNVKQHA DQTVNEDSEG RDLRQVTSTT KSCREKTELT PDQQTLLHFI MDSYNKQRMP QEITNKILKE EFSAEENFLI LTEMATNHVQ VLVEFTKKLP GFQTLDHEDQ IALLKGSAVE AMFLRSAEIF NKKLPSGHSD LLEERIRNSG ISDEYITPMF SFYKSIGELK MTQEEYALLT AIVILSPDRQ YIKDREAVEK LQEPLLDVLQ KLCKIHQPEN PQHFACLLGR LTELRTFNHH HAEMLMSWRV NDHKFT | |
Greater than 90% as determined by SDS-PAGE. |
9971 | |
10 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |