missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl-2-like protein 15 Protein
A DNA sequence encoding the Homo sapiens (Human) Bcl-2-like protein 15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7698-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001010922.1 | |
Bcl-2-like protein 15 Protein | |
Research Use Only | |
BCL2L15 | |
Recombinant Protein | |
E. coli | |
MKSSQTFEEQ TECIVNTLLM DFLSPTLQVA SRNLCCVDEV DSGEPCSFDV AIIAGRLRML GDQFNGELEA SAKNVIAETI KGQTGAILQN TVESLSKTWC AQDSSLAYER AFLAVSVKLL EYMAHIAPEV VGQVAIPMTG MINGNQAIRE FIQGQGGWEN LES | |
Greater than 80% as determined by SDS-PAGE. |
440603 | |
10 μg | |
Not determined. | |
Full Length of BC127719 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |