missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Baculoviral IAP repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Baculoviral IAP repeat-containing protein 8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7945-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_203127.3 | |
Baculoviral IAP repeat-containing protein 1 | |
Research Use Only | |
BIRC8 | |
Recombinant Protein | |
E. coli | |
MTGYEARLIT FGTWMYSVNK EQLARAGFYA IGQEDKVQCF HCGGGLANWK PKEDPWEQHA KWYPGCKYLL EEKGHEYINN IHLTRSLEGA LVQTTKKTPS LTKRISDTIF PNPMLQEAIR MGFDFKDVKK IMEERIQTSG SNYKTLGVLV ADLVSAQKDT TENELNQTSL QREISPEEPL RRLQEEKLCK ICMDRHIAVV FIPCGHLVTC KQCAEAVDRC PMCSMVIDFK QRVFMS | |
Greater than 80% as determined by SDS-PAGE. |
112401 | |
200 μg | |
Not determined. | |
Full Length of BC039318 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |