missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein C-IV Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-IV, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8992-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001637.1 | |
Apolipoprotein C-IV Protein | |
Research Use Only | |
APOC4 | |
Recombinant Protein | |
Yeast | |
CQPEAQEGTL SPPPKLKMSR WSLVRGRMKE LLETVVNRTR DGWQWFWSPS TFRGFMQTYY DDHLRDLGPL TKAWFLESKD SLLKKTHSLC PRLVCGDKDQ G | |
Greater than 90% as determined by SDS-PAGE. |
346 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |