missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apolipoprotein C-IV Protein

A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-IV, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8992-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007421

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001637.1
Apolipoprotein C-IV Protein
Research Use Only
APOC4
Recombinant Protein
Yeast
CQPEAQEGTL SPPPKLKMSR WSLVRGRMKE LLETVVNRTR DGWQWFWSPS TFRGFMQTYY DDHLRDLGPL TKAWFLESKD SLLKKTHSLC PRLVCGDKDQ G
Greater than 90% as determined by SDS-PAGE.
346
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers