missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Annexin A8-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Annexin A8-like protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7711-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060341

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001092315.2
Annexin A8-like protein 1
Research Use Only
ANXA8L2
Recombinant Protein
E. coli
MAWWKAWIEQ EGVTVKSSSH FNPDPDAETL YKAMKGIGVG SQLLSHQAAA FAFPSSALTS VSPWGQQGHL CCNPAGTNEQ AIIDVLTKRS NTQRQQIAKS FKAQFGKDLT ETLKSELSGK FERLIVALMY PPYRYEAKEL HDAMKGSRDD VSSFVDPALA LQDAQDLYAA GEKIRGTDEM KFITILCTRS ATHLLRVKCT QNLHSYFAER LYYAMKGAGT RDGTLIRNIV SRSEIDLNLI KCHFKKMYGK TLSSMIMEDT SGDYKNALLS LVGSDP
Greater than 80% as determined by SDS-PAGE.
728113
500 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers