missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A3 Protein
A DNA sequence encoding the Homo sapiens (Human) Annexin A3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8890-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005130.1 | |
Annexin A3 Protein | |
Research Use Only | |
ANXA3 | |
Recombinant Protein | |
E. coli | |
WVGHRGTVRD YPDFSPSVDA EAIQKAIRGI GTDEKMLISI LTERSNAQRQ LIVKEYQAAY GKELKDDLKG DLSGHFEHLM VALVTPPAVF DAKQLKKSMK GAGTNEDALI EILTTRTSRQ MKDISQAYYT VYKKSLGDDI SSETSGDFRK ALLTLADGRR DESLKVDEHL AKQDAQILYK AGENRWGTDE DKFTEILCLR SFPQLKLTFD EYRNISQKDI VDSIKGELSG HFEDLLLAIV NCVRNTPAFL AERLHRALKG IGTDEFTLNR IMVSRSEIDL LDIRTEFKKH YGYSLYSAIK SDTSGDYEIT LLKICGGD | |
Greater than 90% as determined by SDS-PAGE. |
306 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |