missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A1 Protein
A DNA sequence encoding the Homo sapiens (Human) Annexin A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8765-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000691.1 | |
Annexin A1 Protein | |
Research Use Only | |
ANX1 | |
Recombinant Protein | |
E. coli | |
SEFLKQAWFI ENEEQEYVQT VKSSKGGPGS AVSPYPTFNP SSDVAALHKA IMVKGVDEAT IIDILTKRNN AQRQQIKAAY LQETGKPLDE TLKKALTGHL EEVVLALLKT PAQFDADELR AAMKGLGTDE DTLIEILASR TNKEIRDINR VYREELKRDL AKDITSDTSG DFRNALLSLA KGDRSEDFGV NEDLADSDAR ALYEAGERRK GTDVNVFNTI LTTRSYPQLR RVFQKYTKYS KHDMNKVLDL ELKGDIEKCL TAIVKCATSK PAFFAEKLHQ AMKGVGTRHK ALIRIMVSRS EIDMNDIKAF YQKMYGISLC QAILDETKGD YEKILVALCG GN | |
Greater than 90% as determined by SDS-PAGE. |
301 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |