missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ameloblastin Protein

A DNA sequence encoding the Homo sapiens (Human) Ameloblastin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9909-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058971

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_057603.1
Ameloblastin Protein
Research Use Only
AMBN
Recombinant Protein
Yeast
VPFFPQQSGT PGMASLSLET MRQLGSLQRL NTLSQYSRYG FGKSFNSLWM HGLLPPHSSL PWMRPREHET QQYEYSLPVH PPPLPSQPSL KPQQPGLKPF LQSAAATTNQ ATALKEALQP PIHLGHLPLQ EGELPLVQQQ VAPSDKPPKP ELPGVDFADP QGPSLPGMDF PDPQGPSLPG LDFADPQGST IFQIARLISH GPMPQNKQSP LYPGMLYVPF GANQLNAPAR LGIMSSEEVA GGREDPMAYG AMFPGFGGMR PGFEGMPHNP AMGGDFTLEF DSPVAATKGP ENEEGGAQGS PMPEANPDNL ENPAFLTELE PAPHAGLLAL PKDDIPGLPR SPSGKMKGLP SVTPAAADPL MTPELADVYR TYDADMTTSV DFQEEATMDT TMAPNSLQTS MPGNKAQEPE MMHDAWHFQE P
Greater than 90% as determined by SDS-PAGE.
258
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers