missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 1
A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor-like protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8213-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265222.1 | |
ADP-ribosylation factor-like protein 1 | |
Research Use Only | |
ARL6 | |
Recombinant Protein | |
E. coli | |
GLLDRLSVLL GLKKKEVHVL CLGLDNSGKT TIINKLKPSN AQSQNILPTI GFSIEKFKSS SLSFTVFDMS GQGRYRNLWE HYYKEGQAII FVIDSSDRLR MVVAKEELDT LLNHPDIKHR RIPILFFANK MDLRDAVTSV KVSQLLCLEN IKDKPWHICA SDAIKGEGLQ EGVDWLQDQI QTVKT | |
Greater than 80% as determined by SDS-PAGE. |
84100 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |