missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACY1 / Aminoacylase-1 Protein

A DNA sequence encoding the Homo sapiens (Human) ACY1 / Aminoacylase-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8038-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072311

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_000657.1
ACY1 / Aminoacylase-1 Protein
Research Use Only
ACY1
Recombinant Protein
E. coli
MTSKGPEEEH PSVTLFRQYL RIRTVQPKPD YGAAVAFFEE TARQLGLGCQ KVEVAPGYVV TVLTWPGTNP TLSSILLNSH TDVVPVFKEH WSHDPFEAFK DSEGYIYARG AQDMKCVSIQ YLEAVRRLKV EGHRFPRTIH MTFVPDEEVG GHQGMELFVQ RPEFHALRAG FALDEGIANP TDAFTVFYSE RSPWWVRVTS TGRPGHASRF MEDTAAEKLH KVVNSILAFR EKEWQRLQSN PHLKEGSVTS VNLTKLEGGV AYNVIPATMS ASFDFRVAPD VDFKAFEEQL QSWCQAAGEG VTLEFAQKWM HPQVTPTDDS NPWWAAFSRV CKDMNLTLEP EIMPAATDNR YIRAVGVPAL GFSPMNRTPV LLHDHDERLH EAVFLRGVDI YTRLLPALAS VPALPSDS
Greater than 80% as determined by SDS-PAGE.
95
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers