missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Acetylcholine receptor subunit gamma Protein

A DNA sequence encoding the Homo sapiens (Human) Acetylcholine receptor subunit gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9069-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077561

  • 2650.00€ / 500µg
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_005190.4
Acetylcholine receptor subunit gamma Protein
Research Use Only
CHRNG
Recombinant Protein
Yeast
RNQEERLLAD LMQNYDPNLR PAERDSDVVN VSLKLTLTNL ISLNEREEAL TTNVWIEMQW CDYRLRWDPR DYEGLWVLRV PSTMVWRPDI VLENNVDGVF EVALYCNVLV SPDGCIYWLP PAIFRSACSI SVTYFPFDWQ NCSLIFQSQT YSTNEIDLQL SQEDGQTIEW IFIDPEAFTE NGEWAIQHRP AKMLLDPAAP AQEAGHQKVV FYLLIQRK
Greater than 90% as determined by SDS-PAGE.
1146
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers